Orders LinkContactUsLinkComponent
Omeprazole, Reference Standards
Omeprazole, Reference Standards
Leverancier:   USP
Omeprazole, Reference Standards
Leverancier:  USP
Certificaten
3 Opties Hieronder Beschikbaar
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certificeringen: US Pharmacopea Reference Standard
Documents
Specifications for Product Family
Specificaties
Specificaties
  • Catalog No:
  • 1478505.
  • 1478479.
  • 1478549.
  • Omschrijving:
  • Omeprazole
  • R-Omeprazole
  • Omeprazole Magnesium
  • Synonym(s):
  • 5-Methoxy-2-[(R)-[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]sulFood Ingredientsnyl]-1H-benzimidazole, sodium salt
  • CAS No.:
  • 73590-58-6
  • 161796-77-6
  • 95382-33-5
  • Formule:
  • C₁₇H₁₉N₃O₃S
  • C₁₇H₁₉N₃O₃S
  • C₁₇H₁₉MgN₃O₃S
Specificaties
Product Family Options
Productinformatie
  • Standaard
    Verpakking
    VPE
    Beschikbaarheid
    Price
  • Omeprazole
    USP reference standard
    Vial
    200 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 200 mg
    1 * 200 mg - 405,00 €
    405,00 €
    1 * 200 mg
       405,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    1478505.
    Omschrijving
    Omeprazole
    Synonym(s)
    CAS No.
    73590-58-6
    Formule
    C₁₇H₁₉N₃O₃S
  • 1478479.
    R-Omeprazole
    R-Omeprazole
    USP reference standard
    Vial
    15 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 15 mg
    1 * 15 mg - 1 160,00 €
    1 160,00 €
    1 * 15 mg
       1 160,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    1478479.
    Omschrijving
    R-Omeprazole
    Synonym(s)
    5-Methoxy-2-[(R)-[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]sulFood Ingredientsnyl]-1H-benzimidazole, sodium salt
    CAS No.
    161796-77-6
    Formule
    C₁₇H₁₉N₃O₃S
  • 1478549.
    Omeprazole Magnesium
    Omeprazole Magnesium
    USP reference standard
    Vial
    10 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 10 mg
    1 * 10 mg - 406,00 €
    406,00 €
    1 * 10 mg
       406,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    1478549.
    Omschrijving
    Omeprazole Magnesium
    Synonym(s)
    CAS No.
    95382-33-5
    Formule
    C₁₇H₁₉MgN₃O₃S
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.