Koudebestendige handschoenen, Showa 451
Fournisseur: Showa
Certificats
À propos de cet article
These acrylic/cotton/polyester liners with latex coating on palm and fingertips are ideal for heavy duty handlings in cold environments.
- Abrasion resistant and and very pleasant to wear in cold weather conditions
- Protects the hand in damp environments or against aggressive detergents or alcohols
- Breathable back of hand to reduce perspiration
- Surface provides tactile feel and better grip
- Anatomical shape: Designed for easy movement and continuous wear
- No irritation by seams in contact with the skin
- Low-soil colour
Typical applications include: Agriculture, construction, horticulture, packaging, railways, warehousing & distribution.
Certifications: EN 388:2016 (2241X), EN 511 (010); Cat. II
Spécifications
- Catalog No:
- 112-5837
- 112-5836
- 112-5834
- 112-5835
- Model:
- Showa 451
- Showa 451
- Showa 451
- Showa 451
- Ambidexter/handspecifiek:
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Voor eenmalig gebruik/herbruikbaar:
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Voeringmateriaal:
- Polyester/katoen/acryl
- Polyester/katoen/acryl
- Polyester/katoen/acryl
- Polyester/katoen/acryl
- Afwerking:
- Rough
- Rough
- Rough
- Rough
- Manchetstijl:
- Elastisch
- Elastisch
- Elastisch
- Elastisch
- EN-norm:
- EN 388:2016 (2241X), EN 511 (010)
- EN 388:2016 (2241X), EN 511 (010)
- EN 388:2016 (2241X), EN 511 (010)
- EN 388:2016 (2241X), EN 511 (010)
Spécifications
Frequently Bought Together