Orders LinkContactUsLinkComponent
Detergents reagent solution
Detergents reagent solution
Detergent reagent solution in marked dropping bottle (MDB), Toluidine blue-O chloroform, 100 mL
Catalogus # HACH105932
Leverancier:   Hach
Detergents reagent solution
Catalogus # HACH105932
Leverancier:  Hach
CAS-nummer:  
Detergents reagent solution
Detergents reagent solution
Detergent reagent solution in marked dropping bottle (MDB), Toluidine blue-O chloroform, 100 mL
CAS-nummer:  
Price
47,80 €
Log in voor beschikbaarheid
1 * 100 mL
Product Details & Documents
Used for the determination of detergents (surfactants) by the crystal violet method. For use with storm water test kit.
Verpakking: Reagent is packed in a marked dropping bottle.
Specificaties
Specificaties
  • Chemische methode:
    Toluidine blue-O chloroform
  • VPE:
    100 mL
  • Omschrijving:
    Detergent reagent solution in marked dropping bottle (MDB)