Capsules for microbalances
Silver capsules with capcity: 0,15 ml, height 9 mm and external Ø: 5 mm
Catalogus # LUDI176981126
Leverancier: LUDI
Ga naar:
Product Details & Documents
Made of silver.
- Very good chemical resistance
- Very good temperature resistance
They can be used with all commercially available analysers and microbalances.
Specificaties
- Omschrijving:Silver capsules with capcity: 0,15 ml, height 9 mm and external Ø: 5 mm