Semi-micro cuvette
Semi-micro, 45 mm, 3 ml, PS
Catalogus # 634-8212
Leverancier: Greiner Bio-One
Ga naar:
Product Details & Documents
This semi-micro cuvette is designed for enzymatic determinations.
- Made of crystal clear polystyrene
- Minimal light scatter but high transmission rate
- Thin walls enabling rapid and uniform equilibration
Specificaties
- Type:Semi-micro
- Volume:3 ml
- Venstermateriaal:PS
- Hoogte:45 mm