Order Entry
Belgium
ContactUsLinkComponent
 
Centrifuge tube, thickwall, PC
  BECL355631
Centrifuge tube, thickwall, PC
  BECL355631

 

  • Sluitingstype:
    -
  • Ø×L:
    25×89 mm
  • Uitvoering:
    Open-top, thickwall
  • Capaciteit:
    32 ml
  • Omschrijving:
    For use in High Performance and High Capacity Rotors: JS-24.38 (Part Number 360743) Note: This product replaces Part Number 355637 and 355659.
  • Seq:
    12
  • VPE:
    25

 

 

These open-top, thickwall centrifuge tubes are designed for use with benchtop, high-performance, high-capacity and other ultracentrifuge rotors.