Multi-gas detector, Accuro®
Catalog # DRAE8318392
Supplier: Dräger
Specifications
- Model:Accuro®
- Omschrijving:Multi-gas detector
- VPE:1
Specifications
About this item
Specifically designed for fast measurements with one hand, this gas detector pump allows to use tested tubes under extreme conditions.
Ordering information: Includes hand bellows pump and spares kit in standard leather carrying case.