Orders LinkContactUsLinkComponent
Neutralizing agent, RBS A 375
Neutralizing agent, RBS A 375
Neutralizing agent, RBS A 375, 2.6 (0.5%)
Catalogus # CHEPRBSA375/E123
Leverancier:   CHEMICAL
Neutralizing agent, RBS A 375
Catalogus # CHEPRBSA375/E123
Leverancier:  CHEMICAL
Neutralizing agent, RBS A 375
Neutralizing agent, RBS A 375
Neutralizing agent, RBS A 375, 2.6 (0.5%)
Price
81,90 €
Log in voor beschikbaarheid
1 * 5 L
Product Details & Documents
Improves the cleaning results in washing machines - labwashers.
  • Concentrated solution
  • Surfactant-free
  • Based on citric acid
RBS A 375 solution allows neutralisation and removal of residual alkaline traces and mineral salts from the cleaned items and the internal components of the washing machine. Can also be used in the pre-wash cycle in case of heavy residues. Improves the rinsing cycle and brightens the cleaned items.
 : Available in box of 4 × 5 litres.
Specificaties
  • Omschrijving:
    Neutralizing agent, RBS A 375
  • pH-bereik:
    2.6 (0.5%)