Ultrasonic pipette cleaners
Supplier: BANDELIN-ELECTRONIC
Certificates
About this item
These ultrasonic pipette cleaners are used for gentle cleaning of glass volume-measuring devices and parts with lengths up to 755 mm. having capacity reange from 9 to 18 L.
- Made up of stainless steel
- Kipp handles for easy transport
- Short cleaning times
- Ball valve makes the emptying quickly and easily
Model PR 140 DH consist of optional heating unit have adjustable temperature range from 20 to 80 °C. This heating unit helps to increase the cleaning effect, in the case of grease residue and stubborn soiling.
Delivery: Supplied with stainless steel lid, PE pipette basket (inner dimensions Ø×H: 120×275 mm), universal cleaning concentrates TICKOPUR R 33 (5 L) and TICKOPUR R 27 (1 L).
Specifications
- Catalog No:
- BAND2070
- Model:
- PR 140 DH
- Inhoud:
- 18 L
- Max. uitgangsvermogen:
- 800 W
- b×d×h:
- 150×150×895 mm
Specifications
Frequently Bought Together