Specificaties
- Type antilichaam:Primair
- antigeen symbool:EPHA3
- Clonality:Polyklonaal
- Conjugatie:Cy3®
- Host:Rabbit
- ImmunoChemie:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Kruisadsorptie:No
- Formulier:liquid
- Gen-ID:2042
- Antigeen synoniemen:Tyrosine-protein kinase TYRO4|TYRO4|ETK|Tyrosine-protein kinase receptor ETK1|EPHA3|Ephrin type-A receptor 3|HEK|ETK1|EK4|Eph-like tyrosine kinase 1|Human embryo kinase|hEK4|EPH-like kinase 4
- Modification:phospho-Tyr779
- Opslagbuffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Opslagtemperatuur:Store at –20 °C for 12 months
- Concentratie:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- VPE:100 µl
Specificaties
Over dit artikel
EphA3 is a member of the Ephrin (Eph) family. The Ephrins and Eph-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the Ephrin A (EphA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the Ephrin B (EphB) class, which are transmembrane proteins. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EPHA3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat