Anti-SMYD4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-6108R-A350
: Bioss
- Type antilichaam:Primair
- antigeen symbool:SMYD4
- Clonality:Polyklonaal
- Conjugatie:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemie:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Kruisadsorptie:No
- Formulier:liquid
- Gen-ID:114826
- Antigeen synoniemen:ZMYND21|SMYD4|SMYD4_HUMAN
- Opslagbuffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Opslagtemperatuur:Store at 4°C for 12 months
- Concentratie:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- VPE:100 µl
SMYD4
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SMYD4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit