Order Entry
Belgium
ContactUsLinkComponent
Anti-SYN2 Rabbit Polyclonal Antibody (Cy3®)
Anti-SYN2 Rabbit Polyclonal Antibody (Cy3®)
Catalogus # BOSSBS-3502R-CY3
Leverancier:  Bioss
Anti-SYN2 Rabbit Polyclonal Antibody (Cy3®)
Catalogus # BOSSBS-3502R-CY3
Leverancier:  Bioss

Specificaties

  • Type antilichaam:
    Primair
  • antigeen symbool:
    SYN2
  • Clonality:
    Polyklonaal
  • Conjugatie:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Kruisadsorptie:
    No
  • Formulier:
    liquid
  • Gen-ID:
    6854
  • Antigeen synoniemen:
    Synapsin 2|Synapsin II|Syn2
  • Opslagbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Opslagtemperatuur:
    Store at 4°C for 12 months
  • Concentratie:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • VPE:
    100 µl

Specificaties

Over dit artikel

Synapsin II is a member of the synapsin gene family. Synapsins are neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. This member of the synapsin family is a neuron-specific phosphoprotein that selectively binds to small synaptic vesicles in the presynaptic nerve terminal.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYN2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat