Orders LinkContactUsLinkComponent
Dissolution baskets, QLA
Dissolution baskets, QLA
Suppository basket, plastic
Catalogus # QLAABSKSUP-ST
Leverancier:   QUALITY LAB ACCESSORIES
Dissolution baskets, QLA
Catalogus # QLAABSKSUP-ST
Dissolution baskets, QLA
Dissolution baskets, QLA
Suppository basket, plastic
Price
278,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These are clip style and O-ring push-on style baskets for most dissolution baths. All baskets are laser marked with a unique serial number (except suppository baskets).
  • 40 mesh baskets are made from sintered mesh
  • Different mesh sizes are available
The sintering process essentially creates a welded joint at each overlapping wire junction. Sintered baskets are stronger and last longer. They retain original dimensions and geometry and resist wobble. Most conventional baskets are not sintered and weaken over time. Critical basket geometry is verified on an optical comparator.
Certifications: All baskets are supplied with a certificate of conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications.
Ordering information: Certificate of analysis (CoA) has to be ordered separately.
Specificaties
  • Omschrijving:
    Suppository basket, plastic
  • Seq:
    15
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Basket insertion and removal tool for clip style baskets, black acetal plastic
    Basket insertion and removal tool for clip style baskets, black acetal plastic
    QLAABSKTOOL
    Log in voor beschikbaarheid
    1 * 1 ST - 586,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.