ContactUsLinkComponent
Dual Vespel® ring cross-disk inlet seals
Dual Vespel® ring cross-disk inlet seals
Catalog # REST22084
Supplier:  Restek
undefined
Dual Vespel® ring cross-disk inlet seals
Catalog # REST22084
Supplier:  Restek
Specifications
  • Omschrijving:
    Ring cross-disk inlet seal, Dual Vespel®, Siltek-treated, Ø interior 0,8 mm
  • VPE:
    10
Specifications
About this item
These inlet seals are washerless and leak-tight and ideal for high-flow split applications >500 ml/min.