Capillary GC column cutter, Shortix®
REST23026
: Restek
- Omschrijving:Capillary GC column cutter
- VPE:1
This capillary GC column cutter can be used with 0,25 to 0,53 mm inner diameter and max. 0,78 mm outer diameter tubings.
- Make precise, clean, square cuts with a diamond blade
- Built-in magnifier to verify square cut