Clostridium difficile toxoid B
Catalog # ENZOBMLG1550050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- VPE:50 µG
- Omschrijving:Clostridium difficile toxoid B, 50 µg
- Seq:02
Specifications
About this item
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
- Minimal lot-to-lot variation
- Inactivated by formaldehyde
- Lyophilised for ease of handling and long term stability