Conductivity tester, Eutech™ Expert CTS
Waterproof conductivity tester, Eutech™ Expert CTS
Catalogus # 663-0307
Leverancier: EUTECH
Ga naar:
Product Details & Documents
This waterproof pocket tester is ideal for quick conductivity measurements in hydroponics, aquaculture, agriculture, pools, education, cooling towers, environmental monitoring, water treatment, electroplating and printing.
- Easy to read 4-digit dual-line display with battery life, ready and calibration indicators
- Simultaneous display of conductivity/TDS/salinity and temperature
- Automatic temperature compensation
- Multi-language operational instructions
- Upgraded key pad with intuitive interface
- Symmetrical, leak resistant cap with a fill-line indicator and a wide, stable base
Levering: Supplied with four batteries.
Documents
B-EXPERTPHCTS-E 1016 RevA_WEB
Product documentatie
Specificaties
- Omschrijving:Waterproof conductivity tester, Eutech™ Expert CTS
- TDS - nauwkeurigheid:±2% full scale ±1 LSD
- TDS - resolutie:10 ppm (<2,00 ppt); 0,10 ppt (>2,00 ppt)
- Zoutgehalte - resolutie:0,10 ppt
- Zoutgehalte - nauwkeurigheid:±2% full scale ±1 LSD
- Temperatuurcompensatie:Automatic
- Temperatuurbereik:0...+50,0 °C
- b×d×h:170×45×30 mm
- Temperatuurnauwkeurigheid:±0,5 + 1 LSD °C
- Geleidbaarheid nauwkeurigheid:±2% full scale ±1 LSD
- Voeding:Four batteries (A76 or LR44)
- Geleidbaarheidsbereik:0,00 - 20,00 mS
- TDS - bereik:0,00 - 20,00 ppt
- Cat. No.:663-0307
- Geleidbaarheidsresolutie:10 µS (<2,00 mS); 0,10 mS (>2,00 mS)
- Model:Eutech™ Expert CTS
- Zoutgehalte - bereik:0,0 - 10,0 ppt
- Conformiteit:IP 67
- Temperatuurresolutie:0,1 °C