Tubing cutter, Clean-Cut
REST25069
: Restek
- Omschrijving:Clean-cut tubing cutter
- Seq:01
- VPE:1
This tubing cutter provides burr-free, perpendicular cuts that will not distort the tubing O.D. or close the I.D.
Use for PEEK, PTFE, Tefzel® and other polymeric tubing.