Ice borer
Ice borer, Roestvrij staal 316, 500 mm, 25 mm
Catalogus # SAMP6005A-500
Leverancier: Sampling Systems
Ga naar:
Product Details & Documents
Ice borer specifically designed for ice, wax, and other similar materials.
- Sharp cutting tip
Specificaties
- Ø:25 mm
- Omschrijving:Ice borer
- Lengte:500 mm
- Materiaal:Roestvrij staal 316