0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Injectienaalden
Replacement needle and plunger kit (SGE) for PerkinElmer autosampler syringes
Replacement needle and plunger kit (SGE) for PerkinElmer autosampler syringes
REST24818
:
Restek
Replacement needle and plunger kit (SGE) for PerkinElmer autosampler syringes
REST24818
:
Restek
Meter:
23
Tip type:
Conical needle
Omschrijving:
Replacement needle and plunger kit, 0,5 µl
Lengte:
70 mm
VPE:
1