Jugs, HDPE, Heavy Duty, Nalgene®
NALG642013-0710
- Capaciteit:20 l
- Environmentally Preferable:Lage impact op de fabricage
- Milieu certificering:NAR Certificate of Retirement
- VPE:4
Made of HDPE.
- Good chemical resistance
- Good temperature resistance
- Sturdy