Cylinder holder, wall mounted
REST21333
: Restek
- Type:Cylinder holder, single
- VPE:1
This holder is designed to prevent free-standing gas cylinders from tipping over and injuring personnel.
- Cast aluminum holder can be secured to a wall or the side of a work bench
- Each mount will secure a cylinder 101,6 to 381 mm in diameter