Orders LinkContactUsLinkComponent
Hydrogen flashback arrestor
Hydrogen flashback arrestor
Hydrogen flashback arrestor, brass body, 6,35 mm female N fittings
Catalogus # REST21334
Leverancier:   Restek
Hydrogen flashback arrestor
Catalogus # REST21334
Leverancier:  Restek
Hydrogen flashback arrestor
Hydrogen flashback arrestor
Hydrogen flashback arrestor, brass body, 6,35 mm female N fittings
Price
676,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
This hydrogen flashback arrestor is a special gas safety device, it cuts to stop the flame or reverse flow of gas back up into the equipment or supply line and it prevents the user and equipment from damage or explosions.
  • Valve cuts off gas supply in the event of a flashback
  • Can be reset and reused after flashback has occurred
  • Valve spring is made of 316 stainless steel
Maximum pressure: Hydrogen, 3,5 bar, temperature sensor: polyethylene
Certificeringen: Meets safety requirements of BS6158, EN730, ISO 5175, AS 4603 and are UL Listed 23Y5.
Specificaties
  • Omschrijving:
    Hydrogen flashback arrestor, brass body, 6,35 mm female N fittings