Direct injection liner adapter
Catalogus # REST21303
Leverancier: Restek
Specificaties
- Omschrijving:Direct injection liner adapter for Agilent 5890 packed column GC
- Environmentally Preferable:
- VPE:1
Specificaties
Over dit artikel
This direct injection liner adapter uses standard 1,59 mm capillary nut and ferrules. Can be used with Agilent or Restek direct injection glass inserts or Restek direct injection uniliner liners for an Agilent 5890 packed column GC.
- Convenient wrench pad at base
Levering: Adapter includes 6,35 mm graphite ferrule and stainless steel nut.