Modular gowning bench
Gowning bench, 2510×360×1000 mm
Catalogus # PALBMOD-BENCH-A
Leverancier: Palbam
Ga naar:
Product Details & Documents
The modular gowning bench is an ideal for seating and 'gowning up' in the gown area and allows a flexible design to exactly fit every gown area.
- Made of stainless steel 304L with electro-polished finish
- The seating module reduces the number of legs attached to the floor, making the cleaning process easy and the layout flexible
- A variety of units in various dimensions can be attached in different ways to provide a custom made seating module
Documents
Palbam Class - Catalog 2018
Product documentatie
Palbam Class Cleanroom Furniture
Video 's
Specificaties
- Omschrijving:Gowning bench
- b×d×h:2510×360×1000 mm