GC capillary column connectors kit
Catalog # REST20645
Supplier: Restek
Specifications
- Omschrijving:Capillary inlet adaptor fitting kit, 1/8"
- VPE:1
Specifications
About this item
This split or splitless inlet adaptor fitting is suitable for capillary columns with internal diameter 0,53 mm. Makes column installation easy due to the nut protruding farther from the insulated injection port chamber.
- Accepts standard two-hole capillary ferrules and a standard 1/8 inch nut
- Column insertion depth is the same as the original equipment
- Use recessed gooseneck liners with this adaptor
Ordering information: This kit is supplied with adaptor fitting, capillary nut, stainless steel inlet seal, washer, and one 0,8 mm ID two-hole ferrule.