Digital colony counter, Quebec®
Catalog # REIC13332800
Supplier: REICHERT
Specifications
- Omschrijving:Digital colony counter
- Elektrisch:220 V/50 Hz
- Verlichting:Darkfield
- Vergroting:1,5×
- VPE:1
Specifications
About this item
Digital colony counter delivers easy and accurate count of bacteria colonies with a convenient, built-in digital counter that records and displays the number of counted colonies.
- Glare free illumination for sharp contrast
- Light spreads uniformly on the counting plate for easily distinguishing colonies
- Adjustable culture dish holder for accommodating small culture dishes and rod for easy focusing
- Lens rotates for easy loading of culture dishes
- Built in tilt leg for height and angle adjustment