Open end wrench set
REST20387
: Restek
- Omschrijving:Open end wrench set
- Environmentally Preferable:
- VPE:1
High-quality 1/4×5/16", 3/8"×7/16", 7/16"×1/2", and 1/2"×9/16" wrenches for tightening a wide variety of chromatography fittings.
High-quality 1/4×5/16", 3/8"×7/16", 7/16"×1/2", and 1/2"×9/16" wrenches for tightening a wide variety of chromatography fittings.