ContactUsLinkComponent
VWR® ARISTAR®, Silver Standard Solutions for ICP-MS
VWR® ARISTAR®, Silver Standard Solutions for ICP-MS
Catalogus # 85547.180
Leverancier:  VWR Chemicals
CAS-nummer:  
undefined
VWR® ARISTAR®, Silver Standard Solutions for ICP-MS
Catalogus # 85547.180
Leverancier:  VWR Chemicals
CAS-nummer:  
Specificaties
  • Concentratie:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Verpakking:
    Plastic bottle
  • VPE:
    100 mL
  • Seq:
    117
  • Element:
    Zilver
  • Symbool:
    Ag
Specificaties
Over dit artikel
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Certificeringen: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities