Digital magnifier, Mobilux®
630-2716
- Vergroting:4×; 5×; 6×; 8×; 10×; 12×; 15×
- Omschrijving:Digital magnifier
- Environmentally Preferable:
- VPE:1
The Mobilux® is a portable video magnifier featuring a 4,3" HD TFT LCD screen with anti-glare coating and LED illumination for digital inspection.
- Smaller steps and continuous zoom are also possible
- Three brightness settings, adjustable lines and blinds for precise page orientation
- Various tone settings ensure a comfortable visual experience
It provides time and date and has an automatic shut-off feature after five minutes of non-use and five customisation modes.
A 4 GB SD card stores photos, which are downloadable to a PC or Mac computer through the included USB connection (Windows 10 compatible).
Mobilux® Digital Touch HD software version 2.0 upgrade is available.
: Supplied in a zippered crush-proof protective case is included, along with a cleaning cloth and wrist strap.