Combotester HI 9813 Multi-Parameter Tester (pH/Conductivity/TDS/°C), Handheld
Catalog # SIMEHI9813
Supplier: Industrial Physics
Specifications
- Omschrijving:Multi-parameter tester (pH/conductivity/TDS/°C)
- Model:HI98130
- Geleidbaarheid nauwkeurigheid:±0,2% f.s.
- Geleidbaarheid kalibratie:Automatic, 1 point
- Geleidbaarheidsbereik:0 - 20,00 mS/cm
- Geleidbaarheidsresolutie:0,1 mS/cm
- Aflezing:Dual-display LCD
- Voeding:4 × 1,5 V batteries
- TDS - nauwkeurigheid:±2% f.s.
- TDS - conversiefactor:0,45 - 1,00
- TDS - bereik:0,00 - 10,00 ppt
- TDS - resolutie:0,01 ppt
- Temperatuurcompensatie:pH: automatic; EC/TDS: automatic with ß adjustable from 0,0 to 2,4%/°C
- Gewicht:0,085
- b×d×h:163×40×26
- pH - nauwkeurigheid:±0,05
- pH - kalibratie:Automatic to 1 or 2 point with 2 sets of memorised buffers
- pH - bereik:0,00...14,00
- pH - resolutie:0,01
- VPE:1
Specifications
About this item
The portable combo waterproof tester offer high accuracy pH, conductivity/TDS and temperature measurements in a single tester.
- No more switching between meters for routine measurements
- Waterproof, floating
- Large, easy-to-read, dual-level LCD and automatic shut-off
- pH and EC/TDS readings are automatically compensated for the effects of temperature (ATC)
- EC/TDS conversion factor is user selectable as is the temperature compensation coefficient (ß)
This tester is supplied with a replaceable pH electrode cartridge with an extendable cloth junction as well as an EC/TDS graphite electrode that resists contamination by salts and other substances.
Delivery information: Supplied with pH electrode, electrode removal tool, batteries (4 × 1,5 V) and instructions.