Radius meter
Radius gauge of 20 sheets, 1 - 7 mm
Catalogus # SIMEDI0201
Leverancier: Industrial Physics
Ga naar:
Product Details & Documents
This radius meter provides round edges in outward exposed windows, doors and window frames, with a minimal radius.
Bestelinformatie: Includes 20 sheets.
Documents
TQC Company Video
Video 's
Specificaties
- Meetbereik:1 - 7 mm
- Omschrijving:Radius gauge of 20 sheets