Orders LinkContactUsLinkComponent
Inspection mirror
Inspection mirror
Stainless steel, electrolytically polished inspection mirror, 152×152 mm
Catalogus # MICRQDIST-66P
Leverancier:   MICRONOVA
Inspection mirror
Catalogus # MICRQDIST-66P
Leverancier:  MICRONOVA
Inspection mirror
Inspection mirror
Stainless steel, electrolytically polished inspection mirror, 152×152 mm
Price
491,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Inspection Mirror has a swivel connection allowing it to be attached to stainless steel handle and allows the operator to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned.
  • Highly polished stainless steel
  • A 360° swivel allows optimum flexibility
  • Ideal for hard to reach areas
Bestelinformatie: The optional flashlight attachment is ideal for use in tanks and other low light applications. Rod is not included, but fits to any SSU series handle.
Specificaties
  • Omschrijving:
    Stainless steel, electrolytically polished inspection mirror
  • Lengte:
    152×152 mm
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Flashlight with 6 led, 19-Lumens, batteries 3×AAA (included), 32×97 mm, for use with FLASHH-01 and QDIST series
    Flashlight with 6 led, 19-Lumens, batteries 3×AAA (included), 32×97 mm, for use with FLASHH-01 and QDIST series
    MICRFLASH-01
    Log in voor beschikbaarheid
    1 * 1 ST - 27,10 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Flashlight holder clamp with 360° rotation, black plastic, for FLASH-01 and QDIST series
    Flashlight holder clamp with 360° rotation, black plastic, for FLASH-01 and QDIST series
    MICRFLASHH-01
    Log in voor beschikbaarheid
    1 * 1 ST - 39,70 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.