Orders LinkContactUsLinkComponent
Dissolution paddles, QLA
Dissolution paddles, QLA
Paddle, 419 mm, electropolished 316 stainless steel
Catalogus # QLAAEPD016-02
Leverancier:   QUALITY LAB ACCESSORIES
Dissolution paddles, QLA
Catalogus # QLAAEPD016-02
Dissolution paddles, QLA
Dissolution paddles, QLA
Paddle, 419 mm, electropolished 316 stainless steel
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
QLA paddles are precision manufactured to limit rotational wobble to levels below USP (711) specifications.
  • Made of high quality 316 electropolished stainless steel, PTFE coated over 316 stainless steel or solid fluoropolymer
  • All paddles are laser marked with a unique serial number
The single piece molded Fluoropolymer paddles provide a seamless construction eliminating corrosion and contamination encountered with conventional three-piece assemblies.
Ordering information: Other paddle shaft lengths are also available on request.
Delivery information: All paddles are supplied with a Certificate of Conformance (COC) stating that they are USP compliant and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an additional cost.
Specificaties
  • Omschrijving:
    Paddle, 419 mm, electropolished 316 stainless steel
  • Seq:
    07