Orders LinkContactUsLinkComponent
Test kits, alkalinity (discs), CHECKIT®
Test kits, alkalinity (discs), CHECKIT®
Total alkalinity disc, 20 - 240 mg/l CaCO3, 1 KIT
Catalogus # LOVI146450
Leverancier:   Lovibond
Test kits, alkalinity (discs), CHECKIT®
Catalogus # LOVI146450
Leverancier:  Lovibond
CAS-nummer:  
Test kits, alkalinity (discs), CHECKIT®
Test kits, alkalinity (discs), CHECKIT®
Total alkalinity disc, 20 - 240 mg/l CaCO3, 1 KIT
CAS-nummer:  
Price
29,40 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Delivery information: Supplied with instruction manuals explaining the various stages of analysis.
Specificaties
Specificaties
  • VPE:
    1 KIT
  • Omschrijving:
    Total alkalinity disc
  • Meetbereik:
    20 - 240 mg/l CaCO<sub>3</sub>
Specificaties
Accessories
Productinformatie
  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • CHECKIT® Comparator, path length 13,5 mm
    CHECKIT® Comparator, path length 13,5 mm
    LOVI145000
    1
    Log in voor beschikbaarheid
    1 * 1 ST - 19,30 €
Productinformatie
  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    LOVI145500
    10
    Log in voor beschikbaarheid
    1 * 10 ST - 43,80 €
Productinformatie
  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    LOVI145505
    5
    Log in voor beschikbaarheid
    1 * 5 ST - 25,90 €
Productinformatie
  • Catalogus #
    VPE
    Beschikbaarheid
    Price
  • CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    LOVI145510
    100
    Log in voor beschikbaarheid
    1 * 100 ST - 363,00 €