Turbine spinner for use with Bruker® Spectrometers, SP Wilmad-LabGlass
Catalog # WILMB-PEEK-5
Supplier: SP Industries
Specifications
- Omschrijving:Bruker spinner turbine, PEEK with Viton™ O-rings, 5 mm, suitable for application temperature range: -150... +200 °C
- Materiaal:PEEK
- VPE:1
Specifications
About this item
Manufactured to precision standards that exceed the spectrometer manufacturers requirements.
- Variable temperature operation of -150 to +200 °C
- Weight compatible with room temperature spinners
- Susceptibility matched with Bruker® originals
Can be mixed with other turbines during sample changer operation.