Reversed phase HPLC columns, Kinetex® C18
Supplier: PHENOMENEX
Certificates
About this item
Kinetex C18 are core-shell silica supported 100 Å columns with TMS endcapping for general reversed phase separations.
- Suitable for ultra-high performance for hydrophobic compounds
- High particle density helps create optimal bed structure which reduces band broadening effects of Eddy diffusion
- Ultra-high performance on UHPLC systems
- Reduced diffusion path improves efficiency
Specifications
- Catalog No:
- PHEN00C-4462-E0
- PHEN00D-4475-AN
- PHEN00C-4462-Y0
- PHEN00F-4601-AN
- PHEN00F-4462-Y0
- PHEN00B-4475-AN
- PHEN00G-4601-E0
- PHEN00F-4462-E0
- PHEN00D-4462-E0
- PHEN00F-4601-E0
- PHEN00D-4601-AN
- PHEN00B-4462-AN
- PHEN00B-4601-AN
- PHEN00D-4462-AN
- PHEN00D-4462-Y0
- PHEN00A-4462-AN
- PHEN00B-4515-AN
- PHEN00A-4462-E0
- PHEN00F-4475-AN
- PHEN00F-4462-AN
- Koolstofgehalte:
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- Endcapping:
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Ja
- Max. druk:
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- pH-bereik:
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- 1,5 - 8,5
- Oppervlakte:
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- 200 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
Specifications
Frequently Bought Together