TIC instrument controller, 3 head
TIC instrument controller, with 3 head, RS232/RS485
Catalogus # EDWAD39700000
Leverancier: EDWARDS HIGH VACUUM METTLER
TIC instrument controller, 3 head
Catalogus # EDWAD39700000
Leverancier: EDWARDS HIGH VACUUM METTLER
Ga naar:
Product Details & Documents
TIC Instrument controller is a compact control with a large, clear graphical display, an intuitive user interface and serial communications.
- Automatically recognises and controls active gauges, including APG100 Pirani, convection, thermocouple, strain, inverted magnetron, wide range and active ion gauges
- Enables complete integration into PC and PLC controlled processes, all TIC variants include RS232 and RS485 interface
Windows™-based PC program provides full remote setup, control and data logging functions via the RS232 interface.
Informations de commande: Accessories are sold separately. Contact your Avantor representative for more information.
Specificaties
- Omschrijving:TIC instrument controller, with 3 head, RS232/RS485
- Seq:01
- Cat. No.:EDWAD39700000
- Toegestane omgevingstemperatuur voor bedrijf:0...40 °C (measured underneath TIC)
- Gewicht:45108 kg
- Voedingsvereisten:90 - 264 V AC, 47 - 63 Hz
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.