Bump caps, uvex u-cap premium
Catalogus # 111-1913
Leverancier: UVEX
Specificaties
- Kleur:antraciet
- Omschrijving:Bump cap with short peak
- VPE:1
Specificaties
Over dit artikel
This lightweight bump cap offers maximum wearer comfort and protection against bumps and grazes. The comfortable interior consists of four layers: Shock-absorbing foam and absorbent non-woven material are enclosed in a breathable mesh shell.
- Ergonomic inner shell provides maximum flexibility and optimum adjustment to the shape of the head
- The rip-tape strap allows adjustment to any size
- The curved sides of the bump cap ensure it can be safely and comfortably combined with ear defenders
- Reflective piping around the peak of the bump cap and textile components enhance safety and visibility
- A moulded plastic shell provides extra protection behind the head
Colour: Anthracite
Certificeringen: EN 812