Spanner, open end, SGE
Spanner, 3/16"×1/4" AF, Roestvrij staal, 3/16" and 1/4"
Catalog # 568-0686
Supplier: Trajan Scientific and Medical
Jump to:
Product Details & Documents
Spanner sizes and uses: 3/16" for use with mini unions, 1/4" for use with 1/16" nuts, 5/16" for use with micro control valves.
Specifications
- Omschrijving:Spanner, 3/16"×1/4" AF
- Materiaal:Roestvrij staal
- Gebruik met type kop:3/16" and 1/4"