Racks for microtubes and cryo vials
KOEH2-1642
- Aant. gaten:8x8
- Kleur:rood/wit
- Omschrijving:Compartment size: 12,7x12,7 mm
- b×d×h:143×143×34 mm
- VPE:1
PP rack, square, easy to mount. For 1,5/2,0 ml mircrocentrifuge tubes and cryogenic vials.
- Alphanumeric index of the holes
- Solid bottom of each well to secure vials in place for easier opening and closing
- Fits perfectly and is stackable even when filled with tubes