Orders LinkContactUsLinkComponent
Tissue embedding system, TES 99
Tissue embedding system, TES 99
Compact tissue embedding system, 360×570×300 mm
Catalogus # MEDTTES99.420
Leverancier:   MEDITE
Tissue embedding system, TES 99
Catalogus # MEDTTES99.420
Leverancier:  MEDITE
Tissue embedding system, TES 99
Tissue embedding system, TES 99
Compact tissue embedding system, 360×570×300 mm
Price
3 240,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
TES 99 allows comfortable, rapid and accurate embedding of tissue specimens in paraffin. Due to its modular design, the separate units can be combined according to your needs and meet the individual technical and personal requirements of any laboratory. This cool unit has a capacity of up to 80 blocks that can be chilled at a temperature adjustable down to −10 °C.
  • Unique 3 modular system for an ergonomical working
  • High variety by combining the modular units
  • Rapid embedding of tissue specimens in paraffin, great for large quantities
  • Optional equipped with new heated forceps, hot only at the tip
The cold light lamp assures an optimum illumination of the work area and the optional magnifying lens is simply clicked onto it. The paraffin flow is infinitely adjustable and operated by hand or foot switch. The different temperatures are independently adjustable and visible by digital control. The dispenser unit is equipped with a programmable timer and includes two connections for our heated forceps as well as forceps wells on the left and right side of the nozzles.
Specificaties
  • Omschrijving:
    Compact tissue embedding system, 360×570×300 mm