Flessen, smalle hals, rond
Catalog # MUEL14.383.01
Supplier: MUELLER KREMPEL
Specifications
- Ø uitw.:61,8 mm
- Schroefdraad:43 mm
- Capaciteit:263 ml
- Hoogte:147,5 mm
- VPE:12
Specifications
About this item
Clear glass bottles, without lid.
- Good chemical and physical properties
- For twist-off lids (lids must be ordered seperately)
- Ideal for packaging beverages and foods