Amberlite® IR-120 (H) ion exchange resin
Catalog # L14285.30
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- VPE:250 g
- Seq:3545
Specifications
About this item
:
Merck Index:14,382
Density:1.28
:GHS07
Storage Temperature:Kamertemperatuur
CAS Number:78922-04-0
MDL Number:MFCD00132707