TE buffer, (20X), pH 8,0, autoclaved
Leverancier: Thermo Fisher Scientific
Over dit artikel
Physical Form: Liquid
MDL No.: MFCD00236359
2 Opties Hieronder Beschikbaar
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00236359
Documents
Specifications for Product Family
Specificaties
Specificaties
- Catalog No:
- J62388.AK
- J62388.AP
- Physical form:
- Liquid
- Liquid
- MDL No.:
- MFCD00236359
- MFCD00236359
Specificaties
Product Family Options
Productinformatie
- ConcentratiepH-waardeVPEBeschikbaarheidPrice
- J62388.AKTE buffer concentrate, autoclavedSpecificaties:
More
Specifications
Cat. No.J62388.AKPhysical formLiquidMDL No.MFCD00236359 - J62388.APTE buffer concentrate, autoclavedSpecificaties:
More
Specifications
Cat. No.J62388.APPhysical formLiquidMDL No.MFCD00236359