TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3
Leverancier: Thermo Fisher Scientific
TRIS-Tricine-SDS running buffer (10X), cathode buffer, pH 8,3
Leverancier: Thermo Fisher Scientific
Certificaten
Over dit artikel
Physical Form: Liquid
Applications: For TRIS-Tricine SDS gel electrophoresis
2 Opties Hieronder Beschikbaar
Product Details & Documents
Physical Form: Liquid
Applications: For TRIS-Tricine SDS gel electrophoresis
Documents
Specifications for Product Family
Specificaties
Specificaties
- Catalog No:
- J60992.AP
- J60992.K2
- Physical form:
- Liquid
- Liquid
Specificaties
Product Family Options
Productinformatie
- pH-waardeVPEBeschikbaarheidPrice
- J60992.APTRIS-Tricine-SDS running buffer (10X), cathode bufferTRIS-Tricine-SDS running buffer (10X), cathode buffer8.3500 mLLog in voor beschikbaarheid1 * 500 mL109,00 €Specificaties:
More
Specifications
Cat. No.J60992.APPhysical formLiquid - J60992.K2TRIS-Tricine-SDS running buffer (10X), cathode bufferTRIS-Tricine-SDS running buffer (10X), cathode buffer8.31 LLog in voor beschikbaarheid1 * 1 L201,00 €Specificaties:
More
Specifications
Cat. No.J60992.K2Physical formLiquid