Centrifuge tubes
Supplier: Beckman Coulter
Certificates
About this item
Centrifuge tubes available made from PP, translucent or Ultra-Clear™ material.
Ultra-Clear™ tubes are transparent, enabling easy location of visibly banded or pelleted samples.
Frequently Bought Together