Order Entry
Belgium
ContactUsLinkComponent
Boron hydride-tetrahydrofuran complex 1 mol/l in THF stabilised for synthesis, Sigma-Aldrich®
Boron hydride-tetrahydrofuran complex 1 mol/l in THF stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.41077.0100
Supplier:  Merck
CAS Number:  
Boron hydride-tetrahydrofuran complex 1 mol/l in THF stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.41077.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle
  • VPE:
    100 ml
  • Seq:
    2440

Specifications

Specification Test Results

  • Boiling Point:
    66 °C (1013 hPa) (THF)
  • Flash Point:
    -21.7 °C
  • Explosion Limit:
    1.58 - 12.42 %(V)
  • Density:
    0.876 g/cm3 (25 °C)
  • Ignition Point:
    103 °C
  • Hazard Class:
    4.3
  • Hazard Class Description:
    Dangerous When Wet Material
  • Assay:
    0,8 - 1,2 mol/l

Specification Test Results

About this item

 :
Boiling Pt:66 °C (1013 hPa) (THF)
Density:0.876 g/cm³ (25 °C)
UN:3148
ADR:4.3,I
 :M-Clarity™ quality level = MQ200
Flash Pt:–21.7 °C
 :GHS02
 :GHS05
 :GHS08
 :GHS07
Storage Temperature:Koelkast