Filter papers for breweries
: Ahlstrom
Ahlstrom-Munksjö offers filters for malt analysis and malt filtration according to MEBAK, ASBC and EBC standards.
Grade 41b: Medium fast filtering. Recommended for malt analysis.
Grade 15: Plain, for final attenuation of congress wort.
Grade 470: Plain kieselguhr paper used for fine turbidity.
- Catalog No:
- BINZ120047
- BINZ120048
- BINZ120001
- BINZ120049
- BINZ120050
- BINZ120051
- BINZ120052
- BINZ120053
- BINZ120076
- BINZ120054
- BINZ146279
- BINZ146282
- Nr:
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- 41b
- 41b
- Gewicht:
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 68 g/m²
- 75 g/m²
- 75 g/m²
- Filtratiesnelheid:
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 25 s/10 ml
- 22 s/10 ml
- 22 s/10 ml
Product Family Options
Circles
- NrMaat
Specifications
Cat. No.BINZ120047Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120048Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120001Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120049Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120050Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120051Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120052Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120053Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120076Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 mlSpecifications
Cat. No.BINZ120054Nr15Gewicht68 g/m²Filtratiesnelheid25 s/10 ml
Folded filters
- NrMaat
Specifications
Cat. No.BINZ146279Nr41bGewicht75 g/m²Filtratiesnelheid22 s/10 mlSpecifications
Cat. No.BINZ146282Nr41bGewicht75 g/m²Filtratiesnelheid22 s/10 ml