ContactUsLinkComponent
Cabinets for chemicals, CS-CLASSIC
Cabinets for chemicals, CS-CLASSIC
  135-1938
 :  asecos
Cabinets for chemicals, CS-CLASSIC
  135-1938
 :  asecos
  • Omschrijving:
    Storage cabinet for chemical, without interior equipment, CS-CLASSIC, model CS.195.105.MM.4WD
    Body anthracite grey (RAL 7016) / doors pure white (RAL 9010)
  • b×d×h:
    1055×520×1950 mm
  • Deuren:
    2 folding
  • VPE:
    1
  • Gewicht:
    68
For safe, approved and separate storage of different hazardous materials in one cabinet.
  • Door variants: wing doors with integrated ventilation system for each compartment
  • Natural ventilation at the bottom (vent openings at the cabinet bottom)
  • Connection (NW 75) to a technical exhaust system on the top
  • Double-walled door construction
  • High-quality, scratch-resistant structured paint finish
  • Safety cylinder lock integrated in the turning handle
  • Adjusting aids to compensate for uneven floor
Made in Germany.
Outer body made sturdy all welded construction suitable for highest industrial demands with the attached labelling system.
 : Door colours are available on request. Supplied without interior equipment packages. Interior equipment packages need to be ordered in addition. Please check the website or contact Avantor sales office for further information.
 : Delivery prices available upon request.